Assay Design
Gene Transcripts
- Gene Symbol
- RTS1
- Entrez Gene ID
-
854179
- Gene Name
- Gene Aliases
-
SCS1, YOR014W
- Chromosome Location
-
Chr.XV: 357674 - 359947 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183433.1 |
NP_014657.1 |
1 - 1 |
1946 |
70 |
View Full Product Details