Assay Design
Gene Transcripts
- Gene Symbol
- EXO1
- Entrez Gene ID
-
854198
- Gene Name
- Gene Aliases
-
DHS1, YOR033C
- Chromosome Location
-
Chr.XV: 392416 - 394524 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183452.1 |
NP_014676.1 |
1 - 1 |
1944 |
99 |
View Full Product Details