Assay Design
Gene Transcripts
- Gene Symbol
- TMC1
- Entrez Gene ID
-
854217
- Gene Name
- Gene Aliases
-
YOR052C, YOR29-03
- Chromosome Location
-
Chr.XV: 426772 - 427224 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183471.1 |
NP_014695.1 |
1 - 1 |
352 |
66 |
View Full Product Details