Assay Design
Gene Transcripts
- Gene Symbol
- RGA1
- Entrez Gene ID
-
854294
- Gene Name
- Gene Aliases
-
DBM1, THE1, YOR127W
- Chromosome Location
-
Chr.XV: 561170 - 564193 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183546.1 |
NP_014770.1 |
1 - 1 |
2755 |
71 |
View Full Product Details