Assay Design
Gene Transcripts
- Gene Symbol
- MRPL23
- Entrez Gene ID
-
854321
- Gene Name
- Gene Aliases
-
YOR150W, uL13m
- Chromosome Location
-
Chr.XV: 611999 - 612490 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183569.1 |
NP_014793.1 |
1 - 1 |
355 |
72 |
View Full Product Details