Assay Design
Gene Transcripts
- Gene Symbol
- GLN4
- Entrez Gene ID
-
854339
- Gene Name
- Gene Aliases
-
YOR168W
- Chromosome Location
-
Chr.XV: 649303 - 651732 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183587.3 |
NP_014811.3 |
1 - 1 |
2354 |
104 |
View Full Product Details