Assay Design
Gene Transcripts
- Gene Symbol
- RPB10
- Entrez Gene ID
-
854385
- Gene Name
- Gene Aliases
-
YOR210W
- Chromosome Location
-
Chr.XV: 738320 - 738532 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183629.3 |
NP_014853.3 |
1 - 1 |
45 |
82 |
View Full Product Details