Assay Design
Gene Transcripts
- Gene Symbol
- ODC2
- Entrez Gene ID
-
854397
- Gene Name
- Gene Aliases
-
YOR222W
- Chromosome Location
-
Chr.XV: 758330 - 759253 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183641.1 |
NP_014865.1 |
1 - 1 |
846 |
76 |
View Full Product Details