Assay Design
Gene Transcripts
- Gene Symbol
- RDL2
- Entrez Gene ID
-
854460
- Gene Name
- Gene Aliases
-
AIM42, YOR286W
- Chromosome Location
-
Chr.XV: 850280 - 850729 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183705.3 |
NP_014929.3 |
1 - 1 |
354 |
76 |
View Full Product Details