Assay Design
Gene Transcripts
- Gene Symbol
- HSH49
- Entrez Gene ID
-
854497
- Gene Name
- Gene Aliases
-
YOR319W
- Chromosome Location
-
Chr.XV: 912822 - 913463 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183739.1 |
NP_014964.1 |
1 - 1 |
550 |
80 |
View Full Product Details