Assay Design
Gene Transcripts
- Gene Symbol
- PYK2
- Entrez Gene ID
-
854529
- Gene Name
- Gene Aliases
-
YOR347C
- Chromosome Location
-
Chr.XV: 984942 - 986462 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183767.3 |
NP_014992.3 |
1 - 1 |
1447 |
60 |
View Full Product Details