Assay Design
Gene Transcripts
- Gene Symbol
- FIT3
- Entrez Gene ID
-
854565
- Gene Name
- Gene Aliases
-
YOR383C
- Chromosome Location
-
Chr.XV: 1060441 - 1061055 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183803.3 |
NP_015028.3 |
1 - 1 |
548 |
88 |
View Full Product Details