Assay Design
Gene Transcripts
- Gene Symbol
- MSY1
- Entrez Gene ID
-
856007
- Gene Name
- Gene Aliases
-
YPL097W
- Chromosome Location
-
Chr.XVI: 364952 - 366430 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183911.1 |
NP_015228.1 |
1 - 1 |
951 |
102 |
View Full Product Details