Assay Design
Gene Transcripts
- Gene Symbol
- GDE1
- Entrez Gene ID
-
855994
- Gene Name
- Gene Aliases
-
YPL110C
- Chromosome Location
-
Chr.XVI: 341068 - 344739 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183924.1 |
NP_015215.1 |
1 - 1 |
3551 |
93 |
View Full Product Details