Assay Design
Gene Transcripts
- Gene Symbol
- SET6
- Entrez Gene ID
-
855938
- Gene Name
- Gene Aliases
-
YPL165C
- Chromosome Location
-
Chr.XVI: 237956 - 239077 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183979.1 |
NP_015160.1 |
1 - 1 |
854 |
93 |
View Full Product Details