Assay Design
Gene Transcripts
- Gene Symbol
- MMT2
- Entrez Gene ID
-
855877
- Gene Name
- Gene Aliases
-
MFT2, YPL224C
- Chromosome Location
-
Chr.XVI: 126634 - 128088 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001184038.2 |
NP_015100.2 |
1 - 1 |
1249 |
99 |
View Full Product Details