Assay Design
Gene Transcripts
- Gene Symbol
- BBP1
- Entrez Gene ID
-
855820
- Gene Name
- Gene Aliases
-
YPL255W
- Chromosome Location
-
Chr.XVI: 67725 - 68882 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001184069.1 |
NP_015068.1 |
1 - 1 |
954 |
100 |
View Full Product Details