Assay Design
Gene Transcripts
- Gene Symbol
- ATP15
- Entrez Gene ID
-
855857
- Gene Name
- Gene Aliases
-
YPL271W
- Chromosome Location
-
Chr.XVI: 30079 - 30267 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001184085.1 |
NP_015052.1 |
1 - 1 |
54 |
100 |
View Full Product Details