Assay Design
Gene Transcripts
- Gene Symbol
- TIP41
- Entrez Gene ID
-
856153
- Gene Name
- Gene Aliases
-
YPR040W
- Chromosome Location
-
Chr.XVI: 647305 - 648375 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001184137.1 |
NP_015365.1 |
1 - 1 |
944 |
75 |
View Full Product Details