Assay Design
Gene Transcripts
- Gene Symbol
- DPB2
- Entrez Gene ID
-
856305
- Gene Name
- Gene Aliases
-
YPR175W
- Chromosome Location
-
Chr.XVI: 888974 - 891043 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001184272.1 |
NP_015501.2 |
1 - 1 |
1956 |
103 |
View Full Product Details