Assay Design
Gene Transcripts
- Gene Symbol
- TUB1
- Entrez Gene ID
-
854889
- Gene Name
- Gene Aliases
-
YML085C
- Chromosome Location
-
Chr.XIII: 97941 - 99400 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001182444.1 |
NP_013625.1 |
2 - 2 |
950 |
97 |
View Full Product Details