Assay Design
Gene Transcripts
- Gene Symbol
- RPO26
- Entrez Gene ID
-
856317
- Gene Name
- Gene Aliases
-
RPB6, YPR187W
- Chromosome Location
-
Chr.XVI: 911257 - 911800 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001184284.1 |
NP_015513.1 |
2 - 2 |
350 |
96 |
View Full Product Details