Assay Design



Gene Transcripts

Gene Symbol
RING1
Entrez Gene ID
100153162
Gene Name
ring finger protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 25251507 - 25255526 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190235.1 NP_001177164.1 5 - 6 1034 62
GenBank mRNA AK236439.1 - 5 - 6 1037 62

View Full Product Details