Assay Design



Gene Transcripts

Gene Symbol
SLC1A4
Entrez Gene ID
396600
Gene Name
solute carrier family 1 member 4
Gene Aliases
ASCT1
Chromosome Location
Chr.3: 76938102 - 76970519 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013996100.2 XP_013851554.1 - 2466 71
XM_021087450.1 XP_020943109.1 - 2223 71
GenBank mRNA AY375264.1 - 2 - 3 230 71

View Full Product Details