Assay Design



Gene Transcripts

Gene Symbol
RNH1
Entrez Gene ID
445517
Gene Name
ribonuclease/angiogenin inhibitor 1
Gene Aliases
RI, RNH
Chromosome Location
Chr.2: 281919 - 290522 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021082539.1 XP_020938198.1 - 1558 122
XM_021082540.1 XP_020938199.1 - 1490 122
XM_021082541.1 XP_020938200.1 - 1386 122
XM_021082542.1 XP_020938201.1 - 1439 122
XM_021082543.1 XP_020938202.1 - 2559 122
GenBank mRNA AK392214.1 - 9 - 10 1427 122
M58700.1 - 6 - 7 1044 122

View Full Product Details