Assay Design



Gene Transcripts

Gene Symbol
TFF2
Entrez Gene ID
397420
Gene Name
trefoil factor 2
Gene Aliases
PSP, SML1
Chromosome Location
Chr.13: 205669014 - 205672319 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003358971.3 XP_003359019.2 - 371 65
GenBank mRNA AK344296.1 - 2 - 2 198 65
X51696.1 - - 156 65

View Full Product Details