Assay Design



Gene Transcripts

Gene Symbol
IL9
Entrez Gene ID
100144594
Gene Name
interleukin 9
Gene Aliases
-
Chromosome Location
Chr.2: 138048943 - 138052588 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166043.1 NP_001159515.1 5 - 5 445 131
GenBank mRNA EF055899.2 - 5 - 5 445 131

View Full Product Details