Assay Design



Gene Transcripts

Gene Symbol
PGLYRP1
Entrez Gene ID
397213
Gene Name
peptidoglycan recognition protein 1
Gene Aliases
PGLYRP
Chromosome Location
Chr.6: 52318544 - 52322504 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001260.1 NP_001001260.1 3 - 3 548 72
GenBank mRNA AK399747.1 - 3 - 3 552 72
AY598969.1 - 3 - 3 548 72

View Full Product Details