Assay Design



Gene Transcripts

Gene Symbol
HEBP1
Entrez Gene ID
494567
Gene Name
heme binding protein 1
Gene Aliases
HBP
Chromosome Location
Chr.5: 59493304 - 59523381 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011509.1 NP_001011509.1 3 - 3 351 84
GenBank mRNA AK234172.1 - 3 - 3 572 84
AK235495.1 - 3 - 3 423 84
AK347627.1 - 3 - 3 84
AY610447.1 - 3 - 3 365 84
AY662687.1 - 3 - 3 351 84

View Full Product Details