Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077216.1 | NP_001070684.1 | 6 - 6 | 1351 | 69 |
XM_013995821.2 | XP_013851275.2 | - | 2020 | 69 | |
XM_021093612.1 | XP_020949271.1 | - | 1976 | 69 | |
GenBank mRNA | AK238830.1 | - | 1 - 1 | 1019 | 69 |
AK239411.1 | - | 6 - 6 | 1482 | 69 | |
AK352045.1 | - | 1 - 1 | 550 | 69 | |
DQ972965.1 | - | 6 - 6 | 1351 | 69 |