Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213802.1 | NP_998967.1 | 6 - 6 | 746 | 58 |
XM_005664789.3 | XP_005664846.1 | - | 775 | 58 | |
XM_021093482.1 | XP_020949141.1 | - | 857 | 58 | |
XM_021093483.1 | XP_020949142.1 | - | 1007 | 58 | |
GenBank mRNA | U76256.1 | - | 6 - 6 | 746 | 58 |