Assay Design



Gene Transcripts

Gene Symbol
CYSLTR2
Entrez Gene ID
397243
Gene Name
cysteinyl leukotriene receptor 2
Gene Aliases
CYSLT2
Chromosome Location
Chr.11: 18988665 - 19011015 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214130.2 NP_999295.2 3 - 3 1308 86
GenBank mRNA AB052662.1 - 1 - 1 939 86
AK230761.1 - 3 - 3 1308 86
AK230772.1 - 3 - 3 1308 86

View Full Product Details