Assay Design



Gene Transcripts

Gene Symbol
UTS2
Entrez Gene ID
397268
Gene Name
urotensin 2
Gene Aliases
-
Chromosome Location
Chr.6: 68552889 - 68560827 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214143.1 NP_999308.1 1 - 2 154 67
GenBank mRNA AB063244.1 - 1 - 2 154 67

View Full Product Details