Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214413.1 | NP_999578.1 | 8 - 8 | 1350 | 74 |
GenBank mRNA | AB052256.1 | - | 8 - 8 | 1350 | 74 |
AK391114.1 | - | 9 - 9 | 1381 | 74 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_021094412.1 | XP_020950071.1 | - | 1355 | 74 |