Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214431.2 | NP_999596.1 | 1 - 1 | 47 | 72 |
XM_021083473.1 | XP_020939132.1 | - | 547 | 72 | |
GenBank mRNA | AK234278.1 | - | 1 - 1 | 199 | 72 |
AK395982.1 | - | 1 - 1 | 277 | 72 | |
U92246.1 | - | 1 - 1 | 47 | 72 |