Assay Design



Gene Transcripts

Gene Symbol
BLOC1S3
Entrez Gene ID
100049671
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 3
Gene Aliases
-
Chromosome Location
Chr.6: 51590497 - 51591222 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098586.1 NP_001092056.1 1 - 1 248 144
GenBank mRNA AB271939.1 - 1 - 1 248 144

View Full Product Details