Assay Design



Gene Transcripts

Gene Symbol
RAB33A
Entrez Gene ID
100144495
Gene Name
RAB33A, member RAS oncogene family
Gene Aliases
PR33A
Chromosome Location
Chr.X: 106708402 - 106723803 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123177.1 NP_001116649.1 2 - 2 653 64
XM_005673897.3 XP_005673954.1 - 5055 64
XM_021079378.1 XP_020935037.1 - 5162 64
GenBank mRNA AK390527.1 - 2 - 2 797 64
DQ917627.1 - 2 - 2 653 64

View Full Product Details