Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077217.1 | NP_001070685.1 | 3 - 4 | 377 | 105 |
GenBank mRNA | AK347966.1 | - | 3 - 4 | 515 | 105 |
AK397146.1 | - | 3 - 4 | 515 | 105 | |
DQ972964.1 | - | 3 - 4 | 377 | 105 |