Assay Design



Gene Transcripts

Gene Symbol
CRYBB1
Entrez Gene ID
780429
Gene Name
crystallin beta B1
Gene Aliases
-
Chromosome Location
Chr.14: 44194268 - 44206057 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078681.1 NP_001072149.1 3 - 4 427 68
XM_013982998.2 XP_013838452.1 - 413 68
GenBank mRNA DQ915497.1 - 3 - 4 427 68

View Full Product Details