Assay Design



Gene Transcripts

Gene Symbol
EDN3
Entrez Gene ID
100049663
Gene Name
endothelin 3
Gene Aliases
-
Chromosome Location
Chr.17: 59418667 - 59448382 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098582.2 NP_001092052.1 2 - 3 351 69
XM_021077167.1 XP_020932826.1 - 1382 69
GenBank mRNA AB271922.1 - 2 - 3 351 69
AK236444.1 - 2 - 3 684 69
AK389530.1 - 2 - 3 685 69

View Full Product Details