Assay Design



Gene Transcripts

Gene Symbol
CNPY3
Entrez Gene ID
100126290
Gene Name
canopy FGF signaling regulator 3
Gene Aliases
TNRC5
Chromosome Location
Chr.7: 38014766 - 38025752 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110428.1 NP_001103898.1 5 - 6 764 64
XM_021097865.1 XP_020953524.1 - 780 64
XM_021097866.1 XP_020953525.1 - 888 64

View Full Product Details