Assay Design



Gene Transcripts

Gene Symbol
CSTF1
Entrez Gene ID
100126292
Gene Name
cleavage stimulation factor subunit 1
Gene Aliases
-
Chromosome Location
Chr.17: 56972202 - 56984065 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110430.1 NP_001103900.1 5 - 6 802 115
XM_005673029.3 XP_005673086.1 - 956 115
XM_005673030.3 XP_005673087.1 - 1076 115
XM_021077196.1 XP_020932855.1 - 1126 115
GenBank mRNA AK239052.1 - 5 - 6 801 115
AK396372.1 - 5 - 6 816 115

View Full Product Details