Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114064.2 | NP_001107536.2 | 5 - 6 | 838 | 78 |
XM_021097879.1 | XP_020953538.1 | - | 1121 | 77 | |
XM_021097880.1 | XP_020953539.1 | - | 1047 | 77 | |
GenBank mRNA | AK236966.1 | - | 5 - 6 | 835 | 77 |
AK397732.1 | - | 5 - 6 | 841 | 77 | |
DQ333334.1 | - | 5 - 6 | 838 | 78 |