Assay Design



Gene Transcripts

Gene Symbol
HMGCR
Entrez Gene ID
100144446
Gene Name
3-hydroxy-3-methylglutaryl-CoA reductase
Gene Aliases
HMGR
Chromosome Location
Chr.2: 84380245 - 84401117 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122988.1 NP_001116460.1 16 - 17 2246 71
GenBank mRNA DQ432054.1 - 16 - 17 2246 71
EU726797.1 - 15 - 16 2150 71

View Full Product Details