Assay Design



Gene Transcripts

Gene Symbol
TNXB
Entrez Gene ID
445520
Gene Name
tenascin XB
Gene Aliases
TN-X
Chromosome Location
Chr.7: 24088873 - 24155815 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123204.1 NP_001116676.1 - 7681 104

View Full Product Details