Assay Design



Gene Transcripts

Gene Symbol
TNXB
Entrez Gene ID
445520
Gene Name
tenascin XB
Gene Aliases
TN-X
Chromosome Location
Chr.7: 24088873 - 24155815 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123204.1 NP_001116676.1 - 11776 122
XM_021098327.1 XP_020953986.1 - 14566 122
XM_021098328.1 XP_020953987.1 - 14512 122
XM_021098329.1 XP_020953988.1 - 14512 122
XM_021098330.1 XP_020953989.1 - 14269 122
XM_021098331.1 XP_020953990.1 - 14269 122
XM_021098332.1 XP_020953991.1 - 14239 122
XM_021098333.1 XP_020953992.1 - 14212 122
XM_021098334.1 XP_020953993.1 - 13804 122
XM_021098335.1 XP_020953994.1 - 13618 122
XM_021098336.1 XP_020953995.1 - 13618 122
XM_021098337.1 XP_020953996.1 - 13522 122
XM_021098338.1 XP_020953997.1 - 12544 122
XM_021098339.1 XP_020953998.1 - 8476 122
XM_021098340.1 XP_020953999.1 - 13507 122
GenBank mRNA EU275239.1 - 8 - 9 1571 122
X87243.1 - - 446 122

View Full Product Details