Assay Design



Gene Transcripts

Gene Symbol
PLAU
Entrez Gene ID
396985
Gene Name
plasminogen activator, urokinase
Gene Aliases
uPA
Chromosome Location
Chr.14: 76629313 - 76635173 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213945.1 NP_999110.1 10 - 11 1261 94
GenBank mRNA AK391242.1 - 10 - 11 1277 94
X02724.1 - - 1261 94

View Full Product Details