Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213946.2 | NP_999111.1 | 3 - 4 | 502 | 75 |
XM_013999158.2 | XP_013854612.1 | - | 401 | 75 | |
GenBank mRNA | AK235646.1 | - | 3 - 4 | 75 | |
AK395477.1 | - | 3 - 4 | 539 | 75 | |
DQ472001.1 | - | 3 - 4 | 502 | 75 |