Assay Design



Gene Transcripts

Gene Symbol
IL12A
Entrez Gene ID
397053
Gene Name
interleukin 12A
Gene Aliases
-
Chromosome Location
Chr.13: 99733394 - 99741078 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213993.1 NP_999158.1 3 - 4 489 97
GenBank mRNA KF246517.1 - 3 - 4 382 97
L35765.1 - 3 - 4 489 97

View Full Product Details