Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214050.2 | NP_999215.1 | 5 - 6 | 635 | 79 |
GenBank mRNA | AF188838.3 | - | 5 - 6 | 638 | 79 |
AK235250.1 | - | 5 - 6 | 787 | 79 | |
AK391766.1 | - | 5 - 6 | 662 | 79 | |
AY609655.1 | - | 5 - 6 | 784 | 79 |